452173
Sodium borohydride
granular, ≥98%
Synonym(s):
VenPure® AF, Sodium tetrahydridoborate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
Recommended Products
Assay
≥98%
form
granular
reaction suitability
reagent type: reductant
mp
>300 °C (dec.) (lit.)
SMILES string
[BH4-].[Na+]
InChI
1S/BH4.Na/h1H4;/q-1;+1
InChI key
YOQDYZUWIQVZSF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
Formulated for color and odor removal
Other Notes
This product has been replaced by 452882-ALDRICH | Sodium borohydride powder, ≥98.0%
Legal Information
Product of Rohm & Haas
VenPure is a registered trademark of Ascensus Specialties LLC
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Meganne L Christian et al.
ACS nano, 6(9), 7739-7751 (2012-08-10)
Owing to its high storage capacity (10.8 mass %), sodium borohydride (NaBH(4)) is a promising hydrogen storage material. However, the temperature for hydrogen release is high (>500 °C), and reversibility of the release is unachievable under reasonable conditions. Herein, we
Aldwin Suryo Rahmanto et al.
Free radical biology & medicine, 53(6), 1308-1316 (2012-08-14)
Singlet oxygen ((1)O(2)) is a reactive oxygen species generated during photo-oxidation, inflammation, and via peroxidase-catalyzed reactions (e.g., myeloperoxidase and eosinophil peroxidase). (1)O(2) oxidizes the free amino acids Trp, Tyr, His, Cys, and Met, and those species present on peptides/proteins, with
Marina Baccarin et al.
Materials science & engineering. C, Materials for biological applications, 58, 97-102 (2015-10-20)
A new architecture for a biosensor is proposed using a glassy carbon electrode (GCE) modified with hemoglobin (Hb) and silver nanoparticles (AgNPs) encapsulated in poly(amidoamine) dendrimer (PAMAM). The biosensors were characterized using ultraviolet-visible spectroscopy, ζ-potential and cyclic voltammetry to investigate
Anna V Ogorodnikova et al.
Biochimica et biophysica acta, 1851(9), 1262-1270 (2015-05-27)
Profiling of oxylipins from young maize roots revealed complex patterns of products mainly originating from the combined actions of 9- and 13-lipoxygenases and allene oxide synthase (AOS). A distinctive feature was the high content of the cyclopentenone 10-oxo-11-phytoenoic acid (10-oxo-PEA).
Sylvie Girard et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 72(4), 422-434 (2014-05-29)
Inflammation during pregnancy has devastating consequences for the placenta and fetus. These events are incompletely understood, thereby hampering screening and treatment. The inflammatory profile of villous tissue was studied in pregnancies at high-risk of placental dysfunction and compared to uncomplicated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service