Skip to Content
Merck
All Photos(1)

Key Documents

450669

Sigma-Aldrich

(R)-(+)-4-Isopropyl-5,5-dimethyl-2-oxazolidinone

98%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
R$1,509.00

R$1,509.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
R$1,509.00

About This Item

Empirical Formula (Hill Notation):
C8H15NO2
CAS Number:
Molecular Weight:
157.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

R$1,509.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

optical activity

[α]20/D +47°, c = 1 in H2O

mp

86-87 °C (lit.)

SMILES string

CC(C)[C@H]1NC(=O)OC1(C)C

InChI

1S/C8H15NO2/c1-5(2)6-8(3,4)11-7(10)9-6/h5-6H,1-4H3,(H,9,10)/t6-/m1/s1

InChI key

QICFUFOXXNIZFF-ZCFIWIBFSA-N

Application

Versatile chiral auxiliary for asymmetric synthesis used in diastereoselective Michael additions.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davies, S.G. Sanganee, H.J.
Tetrahedron Asymmetry, 6, 671-671 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service