Skip to Content
Merck
All Photos(1)

Key Documents

448575

Sigma-Aldrich

(−)-Guaiol

97%

Synonym(s):

(3R,6S,10S)-6,10,α,α-Tetramethylbicyclo[5.3.0]dec-1(7)-ene-3-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein:
2047993
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

97%

form

solid

optical activity

[α]20/D −28°, c = 4 in ethanol

mp

91-93 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H]1CC[C@H](CC2=C1CC[C@@H]2C)C(C)(C)O

InChI

1S/C15H26O/c1-10-5-7-12(15(3,4)16)9-14-11(2)6-8-13(10)14/h10-12,16H,5-9H2,1-4H3/t10-,11-,12+/m0/s1

InChI key

TWVJWDMOZJXUID-SDDRHHMPSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Iqbal Choudhary et al.
Journal of natural products, 70(5), 849-852 (2007-03-28)
Microbial transformation of the sesquiterpene (-)-guaiol (1) [1(5)-guaien-11-ol] was investigated using three fungi, Rhizopus stolonifer, Cunninghamella elegans, and Macrophomina phaseolina. Fungal transformation of 1 with Rhizopus stolonifer yielded a hydroxylated product, 1-guaiene-9 beta,11-diol (2). In turn, Cunninghamella elegans afforded two
Xiaohui Yang et al.
Cancer biology & therapy, 19(8), 706-714 (2018-04-04)
(-)-Guaiol, a sesquiterpene alcohol with the guaiane skeleton, has been found in many Chinese medicinal plants and been reported to comprise various guaiane natural products that are well known for their antibacterial activities. Previously, we have shown its antitumor activity
Emma-Tuulia Tolonen et al.
Journal of environmental management, 159, 143-146 (2015-06-13)
The aim of this research was to investigate the effect of magnesium on the removal efficiency of sulphate as gypsum from mine water. The precipitation conditions were simulated with MINEQL + software and the simulation results were compared with the
A L Shreck et al.
Journal of animal science, 93(7), 3613-3622 (2015-10-07)
Two studies were conducted to optimize use of alkaline-treated corn stover and wheat straw and distillers grains as partial corn replacements. In Exp. 1, a finishing experiment used 30 pens (12 steers/pen) of calf-fed steers (initial BW = 374 ±
Jose Manuel Valverde et al.
Physical chemistry chemical physics : PCCP, 17(44), 30162-30176 (2015-10-28)
Thermal decomposition of dolomite in the presence of CO2 in a calcination environment is investigated by means of in situ X-ray diffraction (XRD) and thermogravimetric analysis (TGA). The in situ XRD results suggest that dolomite decomposes directly at a temperature

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service