Skip to Content
Merck
All Photos(1)

Key Documents

446610

Sigma-Aldrich

3,4-Dibromohexane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(Br)CH(Br)C2H5
CAS Number:
Molecular Weight:
243.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.507 (lit.)

bp

39 °C/0.8 mmHg (lit.)

density

1.594 g/mL at 25 °C (lit.)

SMILES string

CCC(Br)C(Br)CC

InChI

1S/C6H12Br2/c1-3-5(7)6(8)4-2/h5-6H,3-4H2,1-2H3

InChI key

VCQBYNRFLHNSKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cathodic elimination reactions of acyclic vicinal dibromides.
Brown OR, et al.
J. Chem. Soc. Perkin Trans. II, 5, 955-963 (1984)
Vibrational spectra of centrosymmetric molecules: Part III. meso-3, 4-dibromohexane and meso-2, 3, 4, 5-tetrabromohexane.
Crowder G A.
Journal of Molecular Structure, 131(3), 245-252 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service