Skip to Content
Merck
All Photos(1)

Key Documents

445924

Sigma-Aldrich

3-Phenyl-1-indanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O
CAS Number:
Molecular Weight:
208.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

148 °C/0.7 mmHg (lit.)

mp

75-78 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C1CC(c2ccccc2)c3ccccc13

InChI

1S/C15H12O/c16-15-10-14(11-6-2-1-3-7-11)12-8-4-5-9-13(12)15/h1-9,14H,10H2

InChI key

SIUOTMYWHGODQX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koltunov KY.
Tetrahedron Letters, 48(32), 5631-5634 (2007)
Asymmetric total synthesis of (+)-indatraline via diastereoselective amination of chiral ethers using chlorosulfonyl isocyanate.
Lee SH, et al.
Tetrahedron, 69(7), 1877-1880 (2013)
Photochemistry of 1-hydroxy-2-indanones: an alternative route to photoenols.
Netto-Ferreira JC, et al.
Journal of the Brazilian Chemical Society, 8(4), 427-431 (1997)
Thermolysis of substituted indenes. Sigmatropic phenyl and hydrogen migrations.
Miller LL and Boyer RF.
Journal of the American Chemical Society, 93(3), 650-656 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service