Skip to Content
Merck
All Photos(1)

Key Documents

444820

Sigma-Aldrich

Fmoc-Asn(Trt)-OH

97%

Synonym(s):

Nα-(9-Fluorenylmethoxycarbonyl)-Nγ-trityl-L-asparagine, Nα-Fmoc-Nγ-trityl-L-asparagine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H32N2O5
CAS Number:
Molecular Weight:
596.67
Beilstein:
4343823
MDL number:
UNSPSC Code:
12352200

Assay

97%

optical activity

[α]25/D −14°, c = 1 in methanol

mp

201-204 °C (lit.)

SMILES string

OC(=O)[C@H](CC(=O)NC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)OCC4c5ccccc5-c6ccccc46

InChI

1S/C38H32N2O5/c41-35(40-38(26-14-4-1-5-15-26,27-16-6-2-7-17-27)28-18-8-3-9-19-28)24-34(36(42)43)39-37(44)45-25-33-31-22-12-10-20-29(31)30-21-11-13-23-32(30)33/h1-23,33-34H,24-25H2,(H,39,44)(H,40,41)(H,42,43)/t34-/m0/s1

InChI key

KJYAFJQCGPUXJY-UMSFTDKQSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service