444154
Yttrium(III) perchlorate solution
40 wt. % in H2O, 99.9% trace metals basis
Synonym(s):
Yttrium triperchlorate
About This Item
vapor pressure
0.12 psi ( 20 °C)
1.68 psi ( 55 °C)
Assay
99.9% trace metals basis
form
liquid
concentration
40 wt. % in H2O
density
1.451 g/mL at 25 °C
SMILES string
[Y+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O
InChI
1S/3ClHO4.Y/c3*2-1(3,4)5;/h3*(H,2,3,4,5);/q;;;+3/p-3
InChI key
IVPOFHHOVYEBFC-UHFFFAOYSA-K
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Liq. 2 - Skin Corr. 1B
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service