Skip to Content
Merck
All Photos(1)

Key Documents

442100

Sigma-Aldrich

Rhodium(II) octanoate, dimer

Rhodium(II) octanoate, dimer
1 of 1 reviewers received a sample product or took part in a promotion

Synonym(s):

Dirhodium tetraoctanoate, Rhodium(II) octanoate dimer, Tetrakis(octanoato)dirhodium

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$1,206.00

R$1,206.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$1,206.00

About This Item

Linear Formula:
[[CH3(CH2)6CO2]2Rh]2
CAS Number:
Molecular Weight:
778.63
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

R$1,206.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

crystalline powder
powder

composition

Rh, 25.0-28.0% gravimetric

reaction suitability

reaction type: C-H Activation
reagent type: catalyst

SMILES string

CCCCCCCC(=O)O[Rh](OC(=O)CCCCCCC)[Rh](OC(=O)CCCCCCC)OC(=O)CCCCCCC

InChI

1S/4C8H16O2.2Rh/c4*1-2-3-4-5-6-7-8(9)10;;/h4*2-7H2,1H3,(H,9,10);;/q;;;;2*+2/p-4

InChI key

FZXFNYFVNKTQSX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Arizona
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Catalyst

    Catalyst for cyclopropanation, was in good conditions

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service