Skip to Content
Merck
All Photos(1)

Key Documents

442089

Sigma-Aldrich

Potassium tetrabromopalladate(II)

Synonym(s):

Dipotassium palladium tetrabromide, Dipotassium tetrabromopalladate, Palladium potassium bromide (PdK2 Br4 ), Potassium tetrabromopalladate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PdBr4
CAS Number:
Molecular Weight:
504.23
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for analytical purposes
form:
crystalline

grade

for analytical purposes

form

crystalline

SMILES string

[K+].[K+].Br[Pd--](Br)(Br)Br

InChI

1S/4BrH.2K.Pd/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

RIURRBMISBBRGH-UHFFFAOYSA-J

Application

Potassium tetrabromopalladate(II) may be used along with (COD)PdCl2 and C3-symmetric N,N′,N′′-tris(2-pyridinylmethyl)-1,3,5-benzenetricarboxamide (L) to prepare Pd6L4-type neutral coordination cages.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoreaction of adsorbed diiodomethane: halide effects of a series of neutral palladium (ii) coordination cages.
Noh TH, et al.
Dalton Transactions, 45(23), 9574-9581 (2016)
Potassium tetrabromopalladate (II).
Martin DS, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 31(10), 2538-2539 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service