Skip to Content
Merck
All Photos(1)

Key Documents

441910

Sigma-Aldrich

6-Bromo-2-benzothiazolinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4BrNOS
CAS Number:
Molecular Weight:
230.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

229-233 °C (lit.)

SMILES string

Brc1ccc2NC(=O)Sc2c1

InChI

1S/C7H4BrNOS/c8-4-1-2-5-6(3-4)11-7(10)9-5/h1-3H,(H,9,10)

InChI key

HECJMTPEVWQFCY-UHFFFAOYSA-N

General description

Synthesis of 6-bromo-2-benzothiazolinone has been reported.[1] Density of 6-bromo-2-benzothiazolinone is 1.7496 g/cm3 at 25°C.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3-Substituted 6-Bromo-2-benzothiazolinones and Their Antialgal and Plant Growth Regulating Activity.
Sidoova E, et al.
Chemical Papers, 46(2), 112-115 (1992)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 2, 136-136 (2015)
K Král'ová et al.
General physiology and biophysics, 12(5), 421-427 (1993-10-01)
2-Alkylthio-6-aminobenzothiazoles and their 6-N-substituted derivatives 3-(2-alkylthio-6-benzothiazolylaminomethyl)-2-benzothiazolineth iones and 3-(2-alkylthio-6-benzothiazolylaminomethyl)-6-bromo-2-benzothia zolinones inhibit photosynthetic processes in spinach chloroplasts and the chlorophyll production in Chlorella vulgaris. The inhibitory activity depends on the alkyl chain length of the thioalkyl substituent. The site of action

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service