441325
Benzene-D6
99.6 atom % D
Synonym(s):
Hexadeuterobenzene
About This Item
Recommended Products
isotopic purity
99.6 atom % D
packaging
pk of 10 × 0.75 mL
refractive index
n20/D 1.497 (lit.)
bp
79.1 °C/760 mmHg (lit.)
mp
6.8 °C (lit.)
density
0.950 g/mL at 25 °C (lit.)
mass shift
M+6
SMILES string
[2H]c1c([2H])c([2H])c([2H])c([2H])c1[2H]
InChI
1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D
InChI key
UHOVQNZJYSORNB-MZWXYZOWSA-N
Looking for similar products? Visit Product Comparison Guide
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1
Target Organs
Blood
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
12.2 °F - closed cup
Flash Point(C)
-11 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service