Skip to Content
Merck
All Photos(1)

Key Documents

438618

Sigma-Aldrich

3-Cyano-4-methylcoumarin

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7NO2
CAS Number:
Molecular Weight:
185.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

192-195 °C (dec.) (lit.)

functional group

ester
nitrile

SMILES string

CC1=C(C#N)C(=O)Oc2ccccc12

InChI

1S/C11H7NO2/c1-7-8-4-2-3-5-10(8)14-11(13)9(7)6-12/h2-5H,1H3

InChI key

FFBLBFMTKGSSPY-UHFFFAOYSA-N

General description

3-Cyano-4-methylcoumarin is a 3-substituted 4-methylcoumarin derivative. Its structural properties, vibrational frequencies and electronic spectra has been studied by density functional theory (DFT).[1] It has been reported to be formed during the Knoevenagel condensation of o-hydroxyacetophenone with ethyl cyanoacetate catalyzed by calcined Mg-Al hydrotalcite.[2] Synthesis of 3-cyano-4-methylcoumarin, via condensation of o-hydroxyacetophenone with ethyl cyanoacetate in the presence of sodium ethylate, has been reported.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Synthesis of Some 3-Substituted-4-methylcoumarins1.
Schroeder CH and Link KP.
Journal of the American Chemical Society, 75(8), 1886-1888 (1953)
N Udaya Sri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 728-736 (2012-08-16)
The vibrational and electronic spectra of 3-cyano-4-methylcoumarin (3C4MC) are reported and discussed. In this work the structural properties, vibrational frequencies and electronic spectra of 3C4MC have been investigated extensively using density functional theory (DFT) employing B3LYP exchange correlation with the
One-pot synthesis of coumarins. Catalysis by the solid base, calcined Mg-Al hydrotalcite.
Ramani A, et al.
Green Chemistry, 1(3), 163-165 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service