Skip to Content
Merck
All Photos(1)

Key Documents

438456

Sigma-Aldrich

Sodium azide

≥99.99% trace metals basis

Synonym(s):

Hydrazoic acid sodium salt, NSC 3072

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaN3
CAS Number:
Molecular Weight:
65.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.99% trace metals basis
form:
solid
solubility:
soluble 65 g/L at 20 °C (completely)

Assay

≥99.99% trace metals basis

form

solid

reaction suitability

reaction type: click chemistry

solubility

soluble 65 g/L at 20 °C (completely)

SMILES string

[Na]N=[N+]=[N-]

InChI

1S/N3.Na/c1-3-2;/q-1;+1

InChI key

PXIPVTKHYLBLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Efficient 1,4-addition of azide to a,ß-unsaturated aldehydes in the presence of tertiary amines.

Catalyst for:
  • Oxidative decarboxylation
  • Michael addition reactions

Reagent for synthesis of
  • Blue fluorescent copolymers
  • Metal phosphonates
  • Arenes via aminations

Involved in regioselective synthesis of prianosin B

suggested gloves for splash protection

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Brain

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 37, 1027-1027 (2007)
H. Chen, et al.,
Macromolecules, 43, 3613-3623 (2010)
Xiaohua Sun et al.
Organic letters, 9(22), 4495-4498 (2007-10-04)
A novel intermolecular cross-double-Michael addition between nitro and carbonyl activated olefins has been developed through Lewis base catalysis. The reaction took place with a large group of beta-alkyl nitroalkenes and alpha,beta-unsaturated ketone/esters, producing an allylic nitro compound in good to
Y. Wada, et al.,
Tetrahedron, 65, 1059-1062 (2009)
Deng-Ke Cao et al.
Inorganic chemistry, 44(10), 3599-3604 (2005-05-10)
Based on the [hydroxy(4-pyridyl)methyl]phosphonate ligand, three compounds with formula Ni{(4-C(5)H(4)N)CH(OH)PO(3)}(H(2)O) (1), Cd{(4-C(5)H(4)N)CH(OH)PO(3)}(H(2)O) (2), and Gd{(4-C(5)H(4)N)CH(OH)P(OH)O(2)}(3).6H(2)O (3) have been synthesized under hydrothermal conditions. The crystal data for 1 are as follows: orthorhombic, space group Pbca, a = 8.7980(13) A, b =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service