Skip to Content
Merck
All Photos(1)

Key Documents

43457

Sigma-Aldrich

(−)-O,O′-Di-pivaloyl-L-tartaric acid

≥98.0% (T)

Synonym(s):

(−)-DPTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O8
CAS Number:
Molecular Weight:
318.32
Beilstein:
2008171
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

optical activity

[α]20/D −23.5±1°, c = 1.7% in dioxane

mp

127-132 °C

functional group

carboxylic acid
ester

SMILES string

CC(C)(C)C(=O)O[C@H]([C@@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O

InChI

1S/C14H22O8/c1-13(2,3)11(19)21-7(9(15)16)8(10(17)18)22-12(20)14(4,5)6/h7-8H,1-6H3,(H,15,16)(H,17,18)/t7-,8-/m1/s1

InChI key

UFHJEZDFEHUYCR-HTQZYQBOSA-N

Application

(−)-O,O′-Di-pivaloyl-L-tartaric acid can be used:
  • As a chiral staple in the synthesis of helical π-conjugated cyclic nanocoils, which are used as nano springs, nano solenoids, and pressure sensors.[1]
  • As an additive in the iridium-catalyzed C-H amidation reactions of arylphosphoryl compounds.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile Bottom-up Approach to Stapled π-Conjugated Helical Scaffolds: Synthesis and Chiroptical Properties of Cyclic o-Phenylene Ethynylene Oligomers
Fuentes N, et al.
Angewandte Chemie (International Edition in English), 124(52), 13213-13217 (2012)
Dual role of carboxylic acid additive: mechanistic studies and implication for the asymmetric C--H amidation
Gwon D, et al.
Tetrahedron, 71(26-27), 4504-4511 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service