Skip to Content
Merck
All Photos(1)

Key Documents

433977

Sigma-Aldrich

5-Nitrofurfuryl alcohol

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO4
CAS Number:
Molecular Weight:
143.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.582 (lit.)

bp

104-105 °C/3 mmHg (lit.)

density

1.283 g/mL at 25 °C (lit.)

SMILES string

OCc1ccc(o1)[N+]([O-])=O

InChI

1S/C5H5NO4/c7-3-4-1-2-5(10-4)6(8)9/h1-2,7H,3H2

InChI key

NDSTUUPEXNCUNW-UHFFFAOYSA-N

Application

5-Nitrofurfuryl alcohol was used to investigate the reduction of some 2-substituted-5-nitrofuran compounds by rhodium complex [Rh(COD)(2-picoline)2](PF6) (COD = 1,5-cyclooctadiene) anchored on poly(4-vinylpyridine).[1] It may be used for the synthesis of 5-nitrofurfuryl bromide and 5-nitrofurfurylamine hydrochloride.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduction of 5-nitrofuran compounds catalyzed by a rhodium complex immobilized on poly (4-vinylpyridine): A relationship with antibacterial activity.
Farkas ME, et al.
Journal of the Chilean Chemical Society, 51(1), 829-835 (2006)
Furan derivatives. XL. Synthesis of 5-nitro-2-substituted furans on the basis of 5-nitrofurfuryl nitrate.
Jurasek A, et al.
Collection of Czechoslovak Chemical Communications, 37(9), 3144-3147 (1972)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service