Skip to Content
Merck
All Photos(1)

Key Documents

432229

Sigma-Aldrich

4-Bromo-2-chloro-1-fluorobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(Cl)F
CAS Number:
Molecular Weight:
209.44
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.553 (lit.)

bp

194 °C (lit.)

density

1.727 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Br)cc1Cl

InChI

1S/C6H3BrClF/c7-4-1-2-6(9)5(8)3-4/h1-3H

InChI key

CJTIWGBQCVYTQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromo-2-chloro-1-fluorobenzene may be used for the preparation of 2-(3-chloro-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark S Jensen et al.
The Journal of organic chemistry, 70(15), 6034-6039 (2005-07-16)
A practical synthesis of 2-[3-(4-fluoro-3-pyridin-3-yl-phenyl)-imidazo[1,2-a]pyrimidin-7-yl]-propan-2-ol (1), an oral GABA(A) alpha(2/3)-selective agonist, is described. The five-step process, which afforded 1 in 40% overall yield, included imidazopyrimidine 2 and pyridine boronic acid 4 as key fragments. The synthesis is highlighted by consecutive
James A Newby et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(1), 263-271 (2013-12-18)
Conducting low-temperature organometallic reactions under continuous flow conditions offers the potential to more accurately control exotherms and thus provide more reproducible and scalable processes. Herein, progress towards this goal with regards to the lithium-halogen exchange/borylation reaction is reported. In addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service