Skip to Content
Merck
All Photos(1)

Key Documents

431451

Sigma-Aldrich

Sodium nitroferricyanide(III) dihydrate

Synonym(s):

Nitroprusside sodium, SNP, Sodium nitroprusside, Sodium pentacyanonitrosylferrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
297.95
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

reagent type: catalyst
core: iron

impurities

≤0.01% insolubles

anion traces

chloride (Cl-): ≤0.02%
sulfate (SO42-): passes test

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O=N[Fe--](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q;;;;;2*-1;2*+1;;

InChI key

OIRZWVYIQXBRFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in medicine hypertensive emergency.

Features and Benefits

Meets A.C.S. reagent specifications.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galen W Heyne et al.
Investigative ophthalmology & visual science, 54(7), 5103-5110 (2013-06-27)
To determine the effect of the nitric oxide donor, sodium nitroprusside (SNP), and the nitric oxide synthase (NOS) inhibitor, L-nitro-arginine-methylester (L-NAME), on IOP, mean arterial pressure (MAP), pupil diameter (PD), refraction (Rfx), aqueous humor formation (AHF), and outflow facility (OF)
Song Guo et al.
Cephalalgia : an international journal of headache, 33(5), 301-307 (2013-02-14)
Sodium nitroprusside (SNP) is a powerful vasodilatory agent that, similarly to glyceryl trinitrate (GTN), releases nitric oxide (NO) but in contrast does not pass the blood-brain barrier. Nevertheless, it has already been used in animal models without any knowledge of
Milos R Filipovic et al.
Journal of medicinal chemistry, 56(4), 1499-1508 (2013-02-20)
Hydrogen sulfide (H2S) has been increasingly recognized as an important signaling molecule that regulates both blood pressure and neuronal activity. Attention has been drawn to its interactions with another gasotransmitter, nitric oxide (NO). Here, we provide evidence that the physiological
Mackram F Eleid et al.
Circulation, 128(12), 1349-1353 (2013-08-21)
Low-gradient severe aortic stenosis with preserved ejection fraction is an increasingly recognized entity, and symptomatic patients may benefit from aortic valve replacement. However, systemic hypertension frequently coexists with low-gradient severe aortic stenosis, which itself may cause elevated left ventricular (LV)
A Moerman et al.
British journal of anaesthesia, 111(4), 619-626 (2013-06-07)
Although both pressure and flow are considered important determinants of regional organ perfusion, the relative importance of each is less established. The aim of the present study was to evaluate the impact of variations in flow, pressure, or both on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service