Skip to Content
Merck
All Photos(1)

Key Documents

43124

Sigma-Aldrich

1-(Diphenylmethyl)piperazine

≥98.0% (NT)

Synonym(s):

1-Benzhydrylpiperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2
CAS Number:
Molecular Weight:
252.35
Beilstein:
222773
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay:
≥98.0% (NT)

Assay

≥98.0% (NT)

impurities

≤5.0% water

SMILES string

C1CN(CCN1)C(c2ccccc2)c3ccccc3

InChI

1S/C17H20N2/c1-3-7-15(8-4-1)17(16-9-5-2-6-10-16)19-13-11-18-12-14-19/h1-10,17-18H,11-14H2

InChI key

NWVNXDKZIQLBNM-UHFFFAOYSA-N

General description

1-(Diphenylmethyl)piperazine is an intermediate during drug synthesis. It has been reported to be formed during the oxidative metabolism of cinnarizine (CZ) [1-(diphenylmethyl)-4-(3-phenyl-2-propenyl)-piperazine] in rat liver microsomes.[1] It reacts with quinone in acetonitrile, followed by oxidation with an alkaline potassium ferricyanide, to afford 4-amino-3,6-di(tert-butyl)-o-benzoquinones.[2]

Application

1-(Diphenylmethyl)piperazine [N-(Diphenylmethyl)piperazine] may be used for the synthesis of 2-nitro-3,4,4-trichloro-1-(propylthio)-1-[4-(diphenylmethyl)piperazin-1-yl]-1,3-butadiene and 2-nitro-3,4,4-trichloro-1-(octadecylthio)-1-[4-(diphenylmethyl)piperazin-1-yl]-1,3-butadiene.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1A - STOT RE 1 - STOT SE 1

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The reactions of polyhalogenated-2-nitro-1, 3-butadiene with alkylthio, thiomorpholine and piperazine derivatives.
Ibis C and Deniz NG.
Indian J. Chem. B, 47(9), 1407-1407 (2008)
M C Dumasia
Xenobiotica; the fate of foreign compounds in biological systems, 32(9), 809-821 (2002-10-25)
1. The in vivo enzymatic Phase I biotransformation of cyclizine (Marezine in the racing greyhound has been shown to proceed via several different pathways. Aromatic and heterocyclic oxidation and the N(4)-demethylation of cyclizine lead to the formation of unconjugated and
Determination of cyclizine and norcyclizine in plasma and urine using gas--liquid chromatography with nitrogen selective detection.
G Land et al.
Journal of chromatography, 222(1), 135-140 (1981-01-02)
D S Griffin et al.
Journal of analytical toxicology, 8(2), 97-99 (1984-03-01)
Gas-liquid chromatography with nitrogen-phosphorus detection was used to analyze blood and urine from a volunteer who ingested 50 mg of cyclizine hydrochloride. A peak blood cyclizine concentration of 69 ng/mL was observed 2 hr after drug administration, and the levels
C S Ananda Kumar et al.
European journal of medicinal chemistry, 44(3), 1223-1229 (2008-11-01)
In order to explore the antiproliferative effect associated with the piperazine framework, several 1-benzhydrylpiperazine derivatives 8(a-d), 9(a-d) and 10(a-h) were synthesized. Variation in the functional group at N-terminal of the piperazine led to three sets of compounds, bearing the sulfonyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service