Skip to Content
Merck
All Photos(2)

Documents

430730

Sigma-Aldrich

2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O5S
CAS Number:
Molecular Weight:
286.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

47-49 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCC2COC(C)(C)O2

InChI

1S/C13H18O5S/c1-10-4-6-12(7-5-10)19(14,15)17-9-11-8-16-13(2,3)18-11/h4-7,11H,8-9H2,1-3H3

InChI key

SRKDUHUULIWXFT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optically active phthalocyanines.
Kobayashi N.
Coordination Chemistry Reviews, 219, 99-123 (2001)
N M Carballeira
Progress in lipid research, 41(6), 437-456 (2002-08-10)
Methoxylated lipids have been reviewed emphasizing the alkylglycerol ethers and fatty acids bearing the methoxy group in the alkyl chain. The literature on methoxylated lipids and their derivatives has been divided into four main groups, namely 2-methoxylated alkyl glycerols, omega-methoxylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service