All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Assay
98%
form
solid
mp
154-156 °C (lit.)
functional group
ester
SMILES string
COC(=O)c1c[nH]cn1
InChI
1S/C5H6N2O2/c1-9-5(8)4-2-6-3-7-4/h2-3H,1H3,(H,6,7)
InChI key
DVLGIQNHKLWSRU-UHFFFAOYSA-N
General description
Methyl 4-imidazolecarboxylate is an alkyl ester of imidazole carboxylic acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
PQQ-sGDH Bioelectrodes Based on Os-Complex Modified Electrodeposition Polymers and Carbon Nanotubes.
Chen X, et al.
Journal of the Electrochemical Society, 161(3), H3058-H3063 (2014)
Visualisation of the local bio-electrocatalytic activity in biofuel cell cathodes by means of redox competition scanning electrochemical microscopy (RC-SECM).
Karnicka K, et al.
Electrochemical Communications, 9(8), 1998-2002 (2007)
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service