Skip to Content
Merck
All Photos(2)

Key Documents

425680

Sigma-Aldrich

Europium(III) trifluoromethanesulfonate

98%

Synonym(s):

Europium(III) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Eu
CAS Number:
Molecular Weight:
599.17
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

core: europium
reagent type: catalyst

SMILES string

[Eu+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Eu/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

TWNOVENTEPVGEJ-UHFFFAOYSA-K

Application

A water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wilko Duprez et al.
PloS one, 10(7), e0133805-e0133805 (2015-08-01)
Antibacterial drugs with novel scaffolds and new mechanisms of action are desperately needed to address the growing problem of antibiotic resistance. The periplasmic oxidative folding system in Gram-negative bacteria represents a possible target for anti-virulence antibacterials. By targeting virulence rather
Atsuko Masuya-Suzuki et al.
ChemistryOpen, 10(1), 46-55 (2021-01-22)
We prepared Ln(III) (Ln=Eu, Gd, and Yb) complexes with a tripodal Schiff base, tris[2-(5-methylsalicylideneimino)ethyl]amine (H3 L) and studied their photophysical properties. Upon ligand excitation, YbL showed Yb(III)-centered luminescence in the near-infrared region. While the overall quantum yield (0.60(1)%) of YbL
The Journal of Organic Chemistry, 59, 3590-3590 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service