Skip to Content
Merck
All Photos(1)

Key Documents

423661

Sigma-Aldrich

Chloroform-d

99.8 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Deuterochloroform

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDCl3
CAS Number:
Molecular Weight:
120.38
Beilstein:
1697633
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.8 atom % D

Assay

≥99.00%

form

liquid

contains

0.03 % (v/v) TMS

packaging

pk of 10 × 1 mL

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.444 (lit.)

bp

60.9 °C (lit.)

mp

−64 °C (lit.)

density

1.500 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]C(Cl)(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Alauddin et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(46), 16479-16493 (2015-09-26)
This work describes the use of conformer-selective laser spectroscopy following supersonic expansion to probe the local folding proclivities of four-membered ring cyclic β-amino acid building blocks. Emphasis is placed on stereochemical effects as well as on the structural changes induced
Manoj Kumar et al.
Cancer research, 74(12), 3271-3281 (2014-04-18)
The preclinical development of peptidyl drugs for cancer treatment is hampered by their poor pharmacologic properties and cell penetrative capabilities in vivo. In this study, we report a nanoparticle-based formulation that overcomes these limitations, illustrating their utility in studies of
Yukiko Tsuji et al.
Plant biotechnology journal, 13(6), 821-832 (2015-01-13)
Bacteria-derived enzymes that can modify specific lignin substructures are potential targets to engineer plants for better biomass processability. The Gram-negative bacterium Sphingobium sp. SYK-6 possesses a Cα-dehydrogenase (LigD) enzyme that has been shown to oxidize the α-hydroxy functionalities in β-O-4-linked
Chong Li et al.
Nature communications, 5, 5709-5709 (2014-12-17)
Photoswitchable fluorescent diarylethenes are promising in molecular optical memory and photonic devices. However, the performance of current diarylethenes is far from satisfactory because of the scarcity of high-speed switching capability and large fluorescence on-off ratio. Here we report a trident
Hui-Ju Chuang et al.
Dalton transactions (Cambridge, England : 2003), 44(2), 598-607 (2014-11-08)
A family of zirconium complexes containing bis-, tri- or tetra-BTP ligands (BTP = benzotriazole phenolate) were synthesized and structurally characterized. Treatment of Zr(O(i)Pr)4((i)PrOH) with 2.0 molar equivalents of 2-(2H-benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol ((C8)BTP-H) or 2-(2H-benzotriazol-2-yl)-4,6-di-tert-butylphenol ((t-Bu)BTP-H) afforded zirconium alkoxide ([((C8)BTP)2Zr(O(i)Pr)2] (1); [((t-Bu)BTP)2Zr(O(i)Pr)2] (2))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service