Skip to Content
Merck
All Photos(2)

Key Documents

422665

Sigma-Aldrich

4,5-Dichlorophthalimide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3Cl2NO2
CAS Number:
Molecular Weight:
216.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

217-219 °C (lit.)

functional group

chloro

SMILES string

Clc1cc2C(=O)NC(=O)c2cc1Cl

InChI

1S/C8H3Cl2NO2/c9-5-1-3-4(2-6(5)10)8(13)11-7(3)12/h1-2H,(H,11,12,13)

InChI key

QJPBDGMPYPSJSF-UHFFFAOYSA-N

General description

4,5-Dichlorophthalimide (5,6-dichloro-1H-isoindole-1,3(2H)-dione) is a phthalimide derivative. It has been synthesized from 5,6-dichloro-1,3-isobenzofurandione and characterized by 1H ,13C-NMR and IR.[1] The dipole moment, polarizability and first-order hyperpolarizability of 4,5-dichlorophthalimide have been investigated using ab initio and density functional theory calculations.[2]

Application

4,5-Dichlorophthalimide (5,6-dichloro-1H-isoindole-1,3(2H)-dione) is suitable as a reactant in the synthesis of 5′-N-(4′′,5′′-dichlorophthaloyl)-3′-azido-2′,3′-dideoxythymidine[3] and 2-amino-4,5-dichlorobenzoic acid.[4]
It may be used in the following studies:
  • As a reactant in the synthesis of 4,5-dichloro-1,2-benzenedicarboxamide.[1]
  • As an additive in the Ir-BICP catalyzed asymmetric hydrogenation of 2,3,3-trimethylindolenine to improve enantioselectivity.[5]
  • As a reactant in the synthesis of octachloro-Cu-phthalocyanine.[6]
  • As a reagent in the synthesis of 7-tert-butylthianthrene-2,3-dicarboxylic imide.[7]
  • As a starting material in the synthesis of N-(3-bromopropyl)-4,5-dichlorophthalimide.[8]
  • As a nucleophilic partner in the phosphine-catalyzed allylic substitution of Morita-Baylis-Hillman acetates to form N-protected β-amino phosphonic acid esters.[9]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HaeIl Park et al.
The Journal of organic chemistry, 71(20), 7892-7894 (2006-09-26)
A series of N-protected beta-amino phosphonic acid esters have been prepared by phosphine-catalyzed allylic substitution of 2-(diethylphosphonyl)-substituted allylic acetates employing 4,5-dichlorophthalimide as nucleophilic partner. These organocatalytic allylic substitutions exhibit exceptionally high levels of regiospecificity by virtue of a tandem S(N)2'-S(N)2'
Conversion of phthalimides to isoindolines by diborane.
8 Strassert CA and Awruch J.
Monatshefte fur Chemie / Chemical Monthly, 137(12), 1499-1503 (2006)
Jan Balzarini et al.
Antiviral chemistry & chemotherapy, 14(3), 139-144 (2003-10-03)
A variety of substituted 5'-N-phthaloyl-3'-azido-2',3'-dideoxythymidine derivatives has been evaluated for their activity against HIV-1, HIV-2 and Moloney murine sarcoma virus (MSV) in cell culture. Most of the 3'-azido-2',3'-dideoxythymidine (AZT, zidovudine) derivatives showed antiviral activity in the lower micromolar concentration range
Elizabeth Sharlow et al.
Probe Reports from the NIH Molecular Libraries Program, 2010 Dec 15 (Updated 2011 24) (2011-10-01)
The unicellular eukaryote
I-Line Sensitive Photoacid Generators Having Thianthrene Skeleton.
Okamura H, et al.
J. Photopolym. Sci. Technol., 17(1), 131-134 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service