422312
Cerium trifluoromethanesulfonate
19-23% Ce basis
Synonym(s):
Cerium triflate
About This Item
form
solid
reaction suitability
core: cerium
reagent type: catalyst
concentration
19-23% Ce
SMILES string
O.[Ce+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
InChI
1S/3CHF3O3S.Ce.H2O/c3*2-1(3,4)8(5,6)7;;/h3*(H,5,6,7);;1H2/q;;;+3;/p-3
InChI key
WETFAJNJKMKTDE-UHFFFAOYSA-K
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service