Skip to Content
Merck
All Photos(1)

Key Documents

421340

Sigma-Aldrich

MERPOL® SE surfactant

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
R$1,098.00

R$1,098.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
R$1,098.00

About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

R$1,098.00


Check Cart for Availability

Request a Bulk Order

form

liquid

refractive index

n20/D 1.454 (lit.)

density

0.97 g/mL at 25 °C (lit.)

HLB

10

General description

Nonionic.

Application

Low foaming wetting agent.

Legal Information

MERPOL is a registered trademark of Stepan Co.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohichi Kawahara et al.
Glia, 64(11), 1938-1961 (2016-07-28)
To differentiate subtypes of microglia (MG), we developed a novel monoclonal antibody, 9F5, against one subtype (type 1) of rat primary MG. The 9F5 showed high selectivity for this cell type in Western blot and immunocytochemical analyses and no cross-reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service