Skip to Content
Merck
All Photos(1)

Key Documents

41740

Sigma-Aldrich

2,2-Dimethylvaleric acid

≥97.0%

Synonym(s):

2,2-Dimethylpentanoic acid, Neoheptanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C(CH3)2COOH
CAS Number:
Molecular Weight:
130.18
Beilstein:
1747703
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.422

density

0.918 g/mL at 20 °C (lit.)

SMILES string

CCCC(C)(C)C(O)=O

InChI

1S/C7H14O2/c1-4-5-7(2,3)6(8)9/h4-5H2,1-3H3,(H,8,9)

InChI key

ZRYCZAWRXHAAPZ-UHFFFAOYSA-N

General description

2,2-Dimethylvaleric acid is a branched-chain-fatty acid. It is a structural analog of di-n-propylacetate (DPA).

Application

2,2-Dimethylvaleric acid may be employed as internal standard for the liquid chromatography-mass spectrometric determination of 2,2-dimethylbutyrate (DMB) in rat plasma.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jorge F Henriques et al.
Aquatic toxicology (Amsterdam, Netherlands), 170, 355-364 (2015-10-21)
Pharmaceuticals are emerging contaminants as their worldwide consumption increases. Fibrates such as gemfibrozil (GEM) are used in human medicine to reduce blood concentrations of cholesterol and triacylglycerol and also are some of the most frequently reported pharmaceuticals in waste waters
J W van der Laan et al.
Pharmacology, biochemistry, and behavior, 13(6), 843-849 (1980-12-01)
An increase in GABA-ergic activity has been implicated in the initiation of quasi-morphine abstinence behavior by di-n-propylacetate (DPA). Two structural analogues of DPA, namely, the branched-chain-fatty acid 2-methyl, 2-ethylcaproic acid and 2,2-dimethylvaleric acid have now been used to study this
J Cunningham et al.
Journal of neurochemistry, 34(1), 197-202 (1980-01-01)
The oxidation of 4-aminobutyric acid (GABA) by nonsynaptosomal mitochondria isolated from rat forebrain and the inhibition of this metabolism by the branched-chain fatty acids 2-methyl-2-ethyl caproate (MEC) and 2.2-dimethyl valerate (DMV) were studied. The rate of GABA oxidation, as measured
Ségolène Siméon et al.
Reproductive toxicology (Elmsford, N.Y.), 93, 219-229 (2020-03-03)
In order to better explain, predict, or extrapolate to humans the developmental toxicity effects of chemicals to zebrafish (Danio rerio) embryos, we developed a physiologically-based pharmacokinetic (PBPK) model designed to predict organ concentrations of neutral or ionizable chemicals, up to
Robert A Parise et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 862(1-2), 168-174 (2007-12-25)
2,2-Dimethylbutyrate (DMB) is a potential treatment for thalassemia and hemoglobinopathies. To facilitate pharmacokinetic evaluation of DMB, we developed an LC-MS assay and quantitated DMB in plasma of rats after an oral dose of 500mg/kg. After acetonitrile protein precipitation, DMB and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service