Skip to Content
Merck
All Photos(1)

Key Documents

416169

Sigma-Aldrich

Trimethylolpropane ethoxylate

average Mn ~170

Synonym(s):

3 arm PEG, 3 arm PEO, Poly(ethylene oxide), 3-arm, hydroxy terminated, Polyethylene glycol trimethylolpropane ether, Polyethylene glycol trimethylolpropane triether, Trimethylolpropane ethoxylate (20/3 EO/OH), Trimethylolpropane ethoxylate (4.6 EO/OH), Trimethylolpropane ethoxylate (4/15 EO/OH)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
R$519.00

R$519.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
R$519.00

About This Item

Linear Formula:
(C2H4O)n(C2H4O)n(C2H4O)nC6H14O
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

R$519.00


Check Cart for Availability

Request a Bulk Order

form

viscous liquid

Quality Level

mol wt

average Mn ~170

refractive index

n20/D 1.478

density

1.11 g/mL at 25 °C

SMILES string

OCC(CO)(CO)CC.OCCO

InChI

1S/C6H14O3.C2H6O2/c1-2-6(3-7,4-8)5-9;3-1-2-4/h7-9H,2-5H2,1H3;3-4H,1-2H2

InChI key

ZSBOTBREDQGUMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

444.2 °F - closed cup

Flash Point(C)

229 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service