Skip to Content
Merck
All Photos(1)

Key Documents

415316

Sigma-Aldrich

(−)-Sparteine

99%

Synonym(s):

(−)-Lupinidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26N2
CAS Number:
Molecular Weight:
234.38
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D −16.5°, c = 10 in ethanol

refractive index

n20/D 1.528 (lit.)

bp

137-138 °C/1 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

C1CCN2C[C@@H]3C[C@@H](CN4CCCC[C@H]34)[C@@H]2C1

InChI

1S/C15H26N2/c1-3-7-16-11-13-9-12(14(16)5-1)10-17-8-4-2-6-15(13)17/h12-15H,1-11H2/t12-,13-,14-,15+/m0/s1

InChI key

SLRCCWJSBJZJBV-ZQDZILKHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Controls asymmetric lithiation/alkylation of prochiral diphenylmethanes and enantioselective deprotonation in preparation of chiral phosphine ligands.
Organocatalyst compexed with copper (II) for the enantioselective syntheses of nitro aldols (Henry reaction).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemical Communications (Cambridge, England), 4006-4006 (2006)
Tetrahedron Letters, 45, 5481-5483 (2004)
Daiju Ichikawa et al.
Biological & pharmaceutical bulletin, 27(9), 1353-1358 (2004-09-02)
Interleukin-12 (IL-12) is a heterodimeric cytokine comprising p40 and p35 subunits produced mainly by monocytes and macrophages, and plays an essential role in the regulation of the differentiation of Th1 cells. Green tea polyphenols exhibit potent anti-oxidative activities and anti-inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service