Skip to Content
Merck
All Photos(2)

Key Documents

414549

Sigma-Aldrich

D-Alanine methyl ester hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CO2CH3·HCl
CAS Number:
Molecular Weight:
139.58
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

optical activity

[α]20/D −8.0°, c = 1.6 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

108-110 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)[C@@H](C)N

InChI

1S/C4H9NO2.ClH/c1-3(5)4(6)7-2;/h3H,5H2,1-2H3;1H/t3-;/m1./s1

InChI key

IYUKFAFDFHZKPI-AENDTGMFSA-N

Application

Building block for the preparation of peptides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Schneider, H. et al.
The Journal of Organic Chemistry, 58, 683-683 (1993)
Davies, J.S.
J. Chem. Soc. Perkin Trans. II, 1225-1225 (1992)
Katarzyna Guzow et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(6), 1133-1140 (2005-03-03)
The photophysical properties of 3-[2-(4-diphenylaminophenyl)benzoxazol-5-yl]alanine methyl ester (1b) and its Boc derivative (1a) were studied in a series of solvents. Its UV-Vis absorption spectra are less sensitive to the solvent polarity than the corresponding fluorescence spectra which show pronounced solvatochromic
B J Bogitsh et al.
The Journal of parasitology, 77(2), 187-193 (1991-04-01)
Cultured Schistosoma mansoni schistosomula of various ages were exposed to several lysosomotropic agents. Weak bases such as chloroquine, ammonium chloride, and acridine orange caused gut swelling upon protonation. The latter compound fluoresced a bright orange indicating the acidic nature of
Peter G W Gettins et al.
The Biochemical journal, 443(1), 65-73 (2011-12-21)
The LRP (low-density lipoprotein receptor-related protein) can bind a wide range of structurally diverse ligands to regions composed of clusters of ~40 residue Ca2+-dependent, disulfide-rich, CRs (complement-like repeats). Whereas lysine residues from the ligands have been implicated in binding, there

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service