Skip to Content
Merck
All Photos(1)

Key Documents

413356

Sigma-Aldrich

Bis[3-(trimethoxysilyl)propyl]amine

technical grade, ≥90%

Synonym(s):

3,3′-Bis(trimethoxysilyl)dipropylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3O)3Si(CH2)3]2NH
CAS Number:
Molecular Weight:
341.55
Beilstein:
9569446
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

≥90%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

152 °C/4 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCCNCCC[Si](OC)(OC)OC)(OC)OC

InChI

1S/C12H31NO6Si2/c1-14-20(15-2,16-3)11-7-9-13-10-8-12-21(17-4,18-5)19-6/h13H,7-12H2,1-6H3

InChI key

TZZGHGKTHXIOMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis[3-(trimethoxysilyl)propyl]amine (BTMSPA) is a bis-amino silane which can be used as a water based coupling agent that promotes the adhesion between different materials to form a hybrid material. The amino groups in the polymer provide coating strength to the modified metal surfaces. The silanol groups bond with the hydroxyls present on the surface of metals which are further cured to give metal-siloxane linkages.[1]

Application

BTMSPA and 1,2-bis(triethoxysilyl)ethane (BTESE) can be used to synthesize an amine modified mesostructured organosilica for potential applications in catalysis and absorbance.[2] Metal finishing may be deviced by coating BTMSPA and vinyltriacetoxysilane which can be used for protection against corrosion.[3]
Bis[3-(trimethoxysilyl)propyl]amine, a silsesquioxane precursor may be used to synthesize hybrid mesostructured organosilica materials.{36}

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced corrosion resistance of AA 2024-T3 and hot-dip galvanized steel using a mixture of bis-[triethoxysilylpropyl] tetrasulfide and bis-[trimethoxysilylpropyl] amine.
Zhu Da and van Ooij WJ
Electrochimica Acta, 49(7), 1113-1125 (2004)
Corrosion protection of metals by water-based silane mixtures of bis-[trimethoxysilylpropyl] amine and vinyltriacetoxysilane.
Zhu D and Van Ooij WJ
Progress in Organic Coatings, 49(1), 42-53 (2004)
Fang Chen et al.
Nanoscale, 9(1), 402-411 (2016-12-08)
Ultrasound is critical in many areas of medicine including obstetrics, oncology, and cardiology with emerging applications in regenerative medicine. However, one critical limitation of ultrasound is the low contrast of target tissue over background. Here, we describe a novel cup-shaped
Bridged amine-functionalized mesoporous organosilica materials from 1,2-bis(triethoxysilyl)ethane and bis((3-trimethoxysilyl)propyl)amine.
Wahab M, et al.
Journal of Solid State Chemistry, 177(10), 3439-3447 (2004)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service