Skip to Content
Merck
All Photos(1)

Key Documents

412171

Sigma-Aldrich

Trimethylolpropane ethoxylate triacrylate

average Mn ~692, contains 100 ppm MEHQ as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHCO2(CH2CH2O)nCH2]3CC2H5
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

mol wt

average Mn ~692

contains

100 ppm MEHQ as inhibitor

refractive index

n20/D 1.471 (lit.)

bp

157 °C (lit.)

density

1.102 g/mL at 25 °C

SMILES string

CCC(COCCOC(=O)C=C)(COCCOC(=O)C=C)COCCOC(=O)C=C

InChI

1S/C21H32O9/c1-5-18(22)28-12-9-25-15-21(8-4,16-26-10-13-29-19(23)6-2)17-27-11-14-30-20(24)7-3/h5-7H,1-3,8-17H2,4H3

InChI key

MTPIZGPBYCHTGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service