Skip to Content
Merck
All Photos(1)

Key Documents

412007

Sigma-Aldrich

2-Chloro-4-methoxyphenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OCH3)OH
CAS Number:
Molecular Weight:
158.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

41-44 °C (lit.)

density

1.153 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

COc1ccc(O)c(Cl)c1

InChI

1S/C7H7ClO2/c1-10-5-2-3-7(9)6(8)4-5/h2-4,9H,1H3

InChI key

GNVRRKLFFYSLGT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Milliken et al.
Applied and environmental microbiology, 70(1), 385-392 (2004-01-09)
The synthesis and degradation of anthropogenic and natural organohalides are the basis of a global halogen cycle. Chlorinated hydroquinone metabolites (CHMs) synthesized by basidiomycete fungi and present in wetland and forest soil are constituents of that cycle. Anaerobic dehalogenating bacteria

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service