Skip to Content
Merck
All Photos(1)

Documents

410136

Sigma-Aldrich

Samarium(III) isopropoxide

99.9% trace metals basis

Synonym(s):

Samarium triisopropoxide, Tris(isopropoxy)samarium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm[OCH(CH3)2]3
CAS Number:
Molecular Weight:
327.62
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder and chunks

reaction suitability

core: samarium
reagent type: catalyst

SMILES string

CC(C)O[Sm](OC(C)C)OC(C)C

InChI

1S/3C3H7O.Sm/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3

InChI key

HJCRVWSKQNDSPZ-UHFFFAOYSA-N

Application

Used with a dinucleating Schiff base ligand in a catalytic asymmetric nitro-Mannich reaction.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

syn-Selective catalytic asymmetric nitro-Mannich reactions using a heterobimetallic Cu-Sm-Schiff base complex.
Shinya Handa et al.
Journal of the American Chemical Society, 129(16), 4900-4901 (2007-03-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service