Skip to Content
Merck
All Photos(1)

Documents

409057

Sigma-Aldrich

Ruthenium(III) nitrosyl chloride hydrate

Synonym(s):

Ruthenium nitroso chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ru(NO)Cl3 · xH2O
CAS Number:
Molecular Weight:
237.44 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

powder

reaction suitability

reagent type: catalyst
core: ruthenium

mp

180 °C (dec.) (lit.)

SMILES string

O.Cl[Ru](Cl)(Cl)N=O

InChI

1S/3ClH.NO.H2O.Ru/c;;;1-2;;/h3*1H;;1H2;/q;;;-1;;+4/p-3

InChI key

GNLMLMMBIPQTRQ-UHFFFAOYSA-K

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service