Skip to Content
Merck
All Photos(3)

Documents

407909

Sigma-Aldrich

N-Z-Ethanolamine

98%

Synonym(s):

2-(Z-Amino)ethanol, Benzyl N-(2-hydroxyethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2NHCO2CH2C6H5
CAS Number:
Molecular Weight:
195.22
Beilstein:
2050891
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

reaction suitability

reagent type: cross-linking reagent

bp

215 °C/15 mmHg (lit.)

mp

58-60 °C (lit.)

functional group

Cbz
amine
hydroxyl

SMILES string

OCCNC(OCC1=CC=CC=C1)=O

InChI

1S/C10H13NO3/c12-7-6-11-10(13)14-8-9-4-2-1-3-5-9/h1-5,12H,6-8H2,(H,11,13)

InChI key

SAGINAGERRNGGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service