Skip to Content
Merck
All Photos(1)

Key Documents

407550

Sigma-Aldrich

Poly(propylene glycol) 4-nonylphenyl ether acrylate

average Mn ~419

Synonym(s):

Nonylphenol polypropylene glycol acrylate, Polypropylene glycol nonylphenyl ether acrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~419

contains

100 ppm MEHQ as inhibitor

refractive index

n20/D 1.49 (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCc1ccc(OCCCOCCCOC(=O)C=C)cc1.CCCCCCCCCc2ccc(OCCCOCCCOCCCOC(=O)C=C)cc2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service