Skip to Content
Merck
All Photos(2)

Key Documents

407402

Sigma-Aldrich

Sodium tetrachloroaluminate

98%

Synonym(s):

Aluminum sodium chloride, Sodium aluminum tetrachloride, Sodium chloroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaAlCl4
CAS Number:
Molecular Weight:
191.78
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

98%

form

powder

reaction suitability

reagent type: catalyst
core: aluminum

mp

185 °C (lit.)

density

2.01 g/mL at 25 °C (lit.)

SMILES string

[Na+].Cl[Al-](Cl)(Cl)Cl

InChI

1S/Al.4ClH.Na/h;4*1H;/q+3;;;;;+1/p-4

InChI key

CMLRNXGMTZKKSR-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service