Skip to Content
Merck
All Photos(1)

Key Documents

406228

Sigma-Aldrich

3-(Trifluoromethyl)pyrazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3F3N2
CAS Number:
Molecular Weight:
136.08
Beilstein:
1564179
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

70 °C/2 mmHg (lit.)

mp

45-47 °C (lit.)

SMILES string

FC(F)(F)c1cc[nH]n1

InChI

1S/C4H3F3N2/c5-4(6,7)3-1-2-8-9-3/h1-2H,(H,8,9)

InChI key

PYXNITNKYBLBMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Trifluoromethyl)pyrazoles is a heterocyclic building block. It undergoes alkylation with alkyl iodides in DMF to afford the N-alkyl pyrazoles. It participates in the synthesis of disubstituted pyrimidines.

Application

3-(Trifluoromethyl)pyrazole may be used in copper-catalyzed pyrazole N-arylation. It may be used in the synthesis of sodium hydridotris(1H-3-trifluoromethylpyrazol-1-yl)borate by heating with sodium borohydride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[A simple method for the direct preparation of O2,3'-cyclo-2'-desoxynucleosides].
G Kowollik et al.
Tetrahedron letters, 44(44), 3863-3865 (1969-09-01)
Mild Conditions for Copper-Catalysed N-Arylation of Pyrazoles.
Cristau HJ, et al.
European Journal of Organic Chemistry, 4, 695-709 (2004)
The effect of the 3-trifluoromethyl substituent in polypyrazolylborato complexes on the iron(II) spin state; X-ray diffraction and absorption and Mossbauer studies.
Cecchi P, et al.
Inorgorganica Chimica Acta, 318(1), 67-76 (2001)
Wei Zhang
Organic letters, 5(7), 1011-1013 (2003-03-28)
[reaction: see text] The fluorous synthesis of disubstituted pyrimidines is carried out by attaching 2,4-dichloro-6-methylpyrimidine with 1H,1H,2H,2H-perfluorodecanethiol. The tagged substrate is substituted with 3-(trifluoromethyl)pyrazole followed by thioether oxidation and tag displacement with amines or thiols. The fluorous chain serves as
Yan Cai et al.
Life sciences, 221, 135-142 (2019-02-08)
Bitter taste receptor (TAS2R) agonists have bronchodilatory potentials. Erythromycin is a ligand of TAS2R10, but its relaxant profile is unknown. This study was performed to understand the relaxant effects of erythromycin and its potential mechanism. Airway resistance was tested by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service