Skip to Content
Merck
All Photos(1)

Key Documents

40253

Sigma-Aldrich

Hydrochloric acid

semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
Pricing and availability is not currently available.

grade

semiconductor grade PURANAL (Honeywell 17823)

vapor density

1.3 (vs air)

vapor pressure

3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)

Assay

37-38%

CofA

specification on request

concentration

30-50%

color

light yellow

bp

>100 °C (lit.)

solubility

H2O: soluble

density

1.18 g/L at 20 °C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Semiconductor grade hydrochloric acid has been characterized by inductively couple plasma mass spectrometry (ICP-MS). The trace detection of V, Cr, As and Se in 20% HCl was determined in the study. [1] It′s efficiency as a microabrasive was studied. [2]

Application

Hydrochloric acid may be used in the formulation of “standard cleaning bath” to purify metallurgical grade silicon (MGS) and polysilicon.[3] Varying concentrations of the acid aided formation of spherical and rod like cellulose nanocrystals.[4] Aqueous mixture of hydrochloric acid and Α-hydroxy acids (lactic, citric, malic, and tartaric acids) may be used to etch a III-V semiconductor (InP). [5]

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical properties of metallurgical-grade silicon in hydrochloric acid.
Kareh KA
Electrochimica Acta, 54(26), 6548-6553 (2009)
Analysis of semiconductor-grade HCl with the ELAN DRC ICP-MS: elimination of chloride-based interferences.
Kishi Y and Kawabata K
Atomic Spectroscopy, 23(5), 165-169 (2002)
Semiconductor Surface-Molecule Interactions, Wet etching of InP by ?-hydroxy acids
Bandaru P and Yablonovitch E
Journal of the Electrochemical Society, 149, G599-G602 (2002)
Charlotte Strandgren et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(8), 3193-3205 (2015-04-17)
Hutchinson-Gilford progeria syndrome (HGPS) is a rare premature aging disorder that is most commonly caused by a de novo point mutation in exon 11 of the LMNA gene, c.1824C>T, which results in an increased production of a truncated form of
Soumya Sinha et al.
The European journal of esthetic dentistry : official journal of the European Academy of Esthetic Dentistry, 8(3), 454-465 (2013-08-21)
The aim of this study was to assess the efficacy of 18% hydrochloric acid and 37% phosphoric acid by an in vivo comparison. Sixty fluorotic permanent maxillary central incisors from 30 patients were divided into 3 categories. The teeth received

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service