Skip to Content
Merck
All Photos(1)

Key Documents

401307

Sigma-Aldrich

Sodium metatitanate

−200 mesh

Synonym(s):

Sodium titanium oxide

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
R$1,177.00

R$1,177.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
R$1,177.00

About This Item

Linear Formula:
Na2Ti3O7
CAS Number:
Molecular Weight:
301.58
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

R$1,177.00


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: titanium
reagent type: catalyst

particle size

−200 mesh

density

3.4 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[O-][Ti](=O)O[Ti](=O)O[Ti]([O-])=O

InChI

1S/2Na.7O.3Ti/q2*+1;;;;;;2*-1;;;

InChI key

GROMGGTZECPEKN-UHFFFAOYSA-N

Application

  • Used as a low-voltage sodium titanium oxide insertion electrode for sodium-ion batteries

Used for preparation of:
  • Titania nanorods and nanowires via a hydrothermal method for use in lithium-ion storage batteries
  • Photocatalytic titanate nanotube thin films via a hydrothermal method
  • Protonated titanate nanotubes as solid acid catalysts
  • Nano-sized supports of ruthenium nanoparticles for catalytic ammonia decomposition

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service