Skip to Content
Merck
All Photos(1)

Key Documents

400033

Sigma-Aldrich

cis-Dichlorobis(pyridine)platinum(II)

97%

Synonym(s):

NSC 129540

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10Cl2N2Pt
CAS Number:
Molecular Weight:
424.18
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: platinum
reagent type: catalyst

mp

282-288 °C (lit.)

SMILES string

Cl[Pt]Cl.[CH]1[CH][CH][N][CH][CH]1.[CH]2[CH][CH][N][CH][CH]2

InChI

1S/2C5H5N.2ClH.Pt/c2*1-2-4-6-5-3-1;;;/h2*1-5H;2*1H;/q;;;;+2/p-2

InChI key

FCJXCLMZOWAKNQ-UHFFFAOYSA-L

Application

Reactant for synthesis of platinum catalysts

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela Horvath et al.
Molecular pharmaceutics, 5(2), 358-363 (2008-02-20)
A novel, water-soluble trans-platinum complex was synthesized by inclusion complexation with beta-cyclodextrin. The complexation was confirmed by (1)H NMR, FT-IR, TGA, and XRD as well as by SEM and EDX. As the precursor complex is not water-soluble, it is difficult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service