399957
Cobalt(II) tetrafluoroborate hexahydrate
99%
Synonym(s):
Cobaltous tetrafluoroborate
About This Item
Recommended Products
Assay
99%
form
crystalline
SMILES string
O.O.O.O.O.O.[Co++].F[B-](F)(F)F.F[B-](F)(F)F
InChI
1S/2BF4.Co.6H2O/c2*2-1(3,4)5;;;;;;;/h;;;6*1H2/q2*-1;+2;;;;;;
InChI key
KQHRHDAVLSRYNA-UHFFFAOYSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Since the preparation of the first organic azide, phenyl azide, by Peter Griess in 1864 this energy-rich and versatile class of compounds has enjoyed considerable interest.
Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service