Skip to Content
Merck
All Photos(1)

Documents

399256

Sigma-Aldrich

(Dimethylphenylsilyl)acetylene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5Si(CH3)2C≡CH
CAS Number:
Molecular Weight:
160.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.508 (lit.)

bp

34 °C/0.07 mmHg (lit.)

density

0.907 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C#C)c1ccccc1

InChI

1S/C10H12Si/c1-4-11(2,3)10-8-6-5-7-9-10/h1,5-9H,2-3H3

InChI key

SSEAMHJAEFEPPK-UHFFFAOYSA-N

General description

(Dimethylphenylsilyl)acetylene is a silylated alkyne. It is reported to participate in the alkynylation of amines catalyzed by Ruthenium complex.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ipaktschi J. and Saidi MR.
Science of Synthesis Knowledge Updates, 4 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service