Skip to Content
Merck
All Photos(1)

Key Documents

391298

Sigma-Aldrich

4-Acetamido-2,2,6,6-tetramethylpiperidine

98%

Synonym(s):

N-(2,2,6,6-Tetramethyl-4-piperidinyl)acetamide, NSC 270075

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O
CAS Number:
Molecular Weight:
198.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

bp

161-163 °C/7 mmHg (lit.)

mp

102-104 °C (lit.)

functional group

amide

SMILES string

CC(=O)NC1CC(C)(C)NC(C)(C)C1

InChI

1S/C11H22N2O/c1-8(14)12-9-6-10(2,3)13-11(4,5)7-9/h9,13H,6-7H2,1-5H3,(H,12,14)

InChI key

HVXYQAREJSKQNE-UHFFFAOYSA-N

General description

4-Acetamido-2,2,6,6-tetramethylpiperidine is a piperidine derivative. Density of 4-acetamido-2,2,6,6-tetramethylpiperidine (N-(2,2,6,6-tetramethyl-4-piperidinyl)acetamide) has been reported.

Application

Reactant for synthesis of:
Selenium and sulfur derivatives with antifungal activity
Oxidants
Alpha and beta-glycosides

Reactant for reaction of nitroxyl radicals with metal carbonyls

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 316-316 (2014)
Jie Hao et al.
International journal of biological macromolecules, 151, 740-746 (2020-02-24)
Oxidized starch (oxStarch) is a major derivative of starch. In present study, 4-acetamide-TEMPO system was firstly applied to prepare specifically oxidized starch, homogeneous 1,4-linked α-D-glucuronan. The impact of oxidant amount, 4-acetamide-TEMPO amount and reaction temperature on the properties of products

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service