Skip to Content
Merck
All Photos(1)

Key Documents

391131

Sigma-Aldrich

3-Azetidinecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
R$936.00

R$936.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
R$936.00

About This Item

Empirical Formula (Hill Notation):
C4H7NO2
CAS Number:
Molecular Weight:
101.10
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

R$936.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

286 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)C1CNC1

InChI

1S/C4H7NO2/c6-4(7)3-1-5-2-3/h3,5H,1-2H2,(H,6,7)

InChI key

GFZWHAAOIVMHOI-UHFFFAOYSA-N

Application

Azetidine and its derivatives are valuable compounds in pharmaceutical[1] and agrochemical[2] research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weed Sci., 38, 315-315 (1990)
G Johnson et al.
Journal of medicinal chemistry, 35(2), 233-241 (1992-01-24)
3-Carboxy-3,4-dehydropyrrolidine was found to bind with affinity equal to that of glycine in a [3H]strychnine binding assay. Simple substitution of the 1-, 2-, 4-, or 5-position resulted in marked loss of affinity. A decline in affinity was also found upon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service