Skip to Content
Merck
All Photos(1)

Key Documents

388130

Sigma-Aldrich

Di(propylene glycol) butyl ether, mixture of isomers

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
R$352.00

R$352.00


Available to ship onApril 09, 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
R$352.00

About This Item

Linear Formula:
CH3(CH2)3OC3H6OC3H6OH
CAS Number:
Molecular Weight:
190.28
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

R$352.00


Available to ship onApril 09, 2025Details


Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.426 (lit.)

bp

222-232 °C (lit.)

density

0.913 g/mL at 25 °C (lit.)

SMILES string

O(C(COCCCC)C)CC(O)C

InChI

1S/C10H22O3/c1-4-5-6-12-8-10(3)13-7-9(2)11/h9-11H,4-8H2,1-3H3

InChI key

CUVLMZNMSPJDON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. ジプロピレングリコールブチルエーテル(cas No.29911-28-2, カタログ番号388130-1L)にBHTは添加されていますでしょうか。 How much BHT is added to dipropylene glycol butyl ether (cas No.29911-28-2, catalog No.388130-1L)?

    1 answer
    1. This product does not contain BHT.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service