Skip to Content
Merck
All Photos(1)

Key Documents

387800

Sigma-Aldrich

5-Chloro-2,4,6-trifluoropyrimidine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4ClF3N2
CAS Number:
Molecular Weight:
168.50
Beilstein:
610168
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.439 (lit.)

bp

116 °C (lit.)

density

1.626 g/mL at 25 °C (lit.)

SMILES string

Fc1nc(F)c(Cl)c(F)n1

InChI

1S/C4ClF3N2/c5-1-2(6)9-4(8)10-3(1)7

InChI key

GOYNRDSJTYLXBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Chloro-2,4,6-trifluoropyrimidine can be used as a starting material to synthesize:
  • 4-azido-5-chloro-2,6-difluoro-pyrimidine by azidation reaction with sodium azide.[1]
  • 5-chloro triphenoxy pyrimidine by reacting with tetraphenoxysilane in the presence of tetra butyl ammonium fluoride (TBAF).[2]
  • 5-chloro-2,6-difluoropyrimidin-4-amine, and 5-chloro-4,6-difluoropyrimidin-2-amine (9:1 ratio) by reacting with ammonia.[3]
  • 5-Chloro-N,N -diethyl-2,6-dipyrimidin-4-amine, by treating with diethylamine and DIPEA.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyfunctionalised pyrimidines and pyrazines from perhalogenated precursors (2008)
An efficient strategy for the synthesis of aryl ethers
Wang T and Love JA
Synthesis, 2007(15), 2237-2239 (2007)
Studies in azide chemistry. Part IX [1]. Investigations involving fluorinated azidopyrimidines and 4-azido-3-chloro-2, 5, 6-trifluoropyridine
Banks RE, et al.
Journal of Fluorine Chemistry, 16(4), 325-338 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service