386251
1-(3-Sulfopropyl)-2-{[3-(3-sulfopropyl)-2(3H)-benzothiazolylidene]methyl}naphtho[1,2-d]thiazolium hydroxide inner salt, triethylammonium salt
Dye content 85 %
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C31H39N3O6S4
CAS Number:
Molecular Weight:
677.92
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
composition
Dye content, 85%
mp
260 °C (dec.) (lit.)
λmax
441 nm
SMILES string
CCN(CC)CC.OS(=O)(=O)CCCN1\C(Sc2ccccc12)=C\c3sc4ccc5ccccc5c4[n+]3CCCS([O-])(=O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service