Skip to Content
Merck
All Photos(1)

Key Documents

380369

Sigma-Aldrich

o-Tolidine dihydrochloride

97%

Synonym(s):

3,3′-Dimethylbenzidine dihydrochloride, 4,4′-Bianisidine dihydrochloride, 4,4′-Diamino-3,3′-dimethylbiphenyl dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H3(CH3)-4-NH2]2 · 2HCl
CAS Number:
Molecular Weight:
285.21
Beilstein:
3713238
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

SMILES string

Cl.Cl.Cc1cc(ccc1N)-c2ccc(N)c(C)c2

InChI

1S/C14H16N2.2ClH/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12;;/h3-8H,15-16H2,1-2H3;2*1H

InChI key

LUKPNZHXJRJBAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sensitive colorimetric reagent for gold and for free chlorine in water.

Warning

May be carcinogenic.

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dyes metabolized to 3,3'-dimethylbenzidine (3,3'-dimethylbenzidine dye class.
Report on carcinogens : carcinogen profiles, 10, 105-106 (2004-08-25)
Zhijun Cao et al.
Biosensors & bioelectronics, 23(3), 348-354 (2007-06-06)
The electrooxidation of o-tolidine (oTD) was investigated via the electrochemical quartz crystal microbalance (EQCM) technique. The formation and breakage of the poorly soluble charge-transfer complex (CTC) occurred during the redox switching of oTD, and the CTC precipitation on and its
3,3'-Dimethylbenzidine.
Report on carcinogens : carcinogen profiles, 10, 104-105 (2004-08-25)
L E Gray et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 20(2), 177-183 (1993-02-01)
In mice and rats, prenatal exposure to the dye Congo red permanently reduces the number of germ cells in male and female offspring. In the current investigation, nine other dyes structurally related to Congo red were examined for developmental testicular
3,3'-Dimethylbenzidine and dyes metabolized to 3,3'-dimethylbenzidine: 3,3'-dimethylbenzidine.
Report on carcinogens : carcinogen profiles, 12, 168-170 (2011-08-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service