Skip to Content
Merck
All Photos(1)

Key Documents

379220

Sigma-Aldrich

(−)-Ambroxide

99%

Synonym(s):

1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.04,9]tridecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28O
CAS Number:
Molecular Weight:
236.39
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D −29°, c = 1 in toluene

mp

74-76 °C (lit.)

SMILES string

CC1(C)CCC[C@@]2(C)[C@H]1CC[C@@]3(C)OCC[C@H]23

InChI

1S/C16H28O/c1-14(2)8-5-9-15(3)12(14)6-10-16(4)13(15)7-11-17-16/h12-13H,5-11H2,1-4H3/t12-,13+,15-,16+/m0/s1

InChI key

YPZUZOLGGMJZJO-LQKXBSAESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ambroxide is a terpenoid, which has vast applications in the perfume industry due to its fixative property and odor. Ambergris, which was originally sourced from sperm whale, has been substituted by synthetic ambroxides.

Application

(−)-Ambroxide can be used:
  • To prepare (+)-sclareolide through C−H oxidation strategy.
  • As a substrate in C(sp3)-H alkylation/arylation studies of ethers.
  • As a substrate in the study of ethereal hydrocarbon hydroperoxidation using singlet O2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymes for Synthetic Biology of Ambroxide-Related Diterpenoid Fragrance Compounds
Biotechnology of Isoprenoids (2015)
Recent developments in natural product synthesis using metal-catalysed C-H bond functionalisation
McMurray L, et al.
Chemical Society Reviews, 40(4), 1885-1898 (2011)
The Combination of Benzaldehyde and Nickel-Catalyzed Photoredox C (sp3)- H Alkylation/Arylation
Zhang L, et al.
Angewandte Chemie (International Edition in English), 58(6), 1823-1827 (2019)
Singlet oxygen-mediated selective C--H bond hydroperoxidation of ethereal hydrocarbons
Sagadevan A, et al.
Nature Communications, 8(1), 1-8 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service